Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Highly Reusable Full-Finger Nylon Glove Liners
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- 100% continuous nylon filament construction is thin and comfortable
- Full finger design offers protection for those with severe glove sensitivities
- Launderable up to 30 times without any signs of shrinkage
- Color-coded hems for easy size identification
- Made in the USA
- Category II for particulates
Specifications
Coating Material | Uncoated |
Color | White |
Disposable | Reusable |
Hand | Ambidextrous |
Material | Nylon |
Product Type | Nylon Glove Liner |
Size | Medium |
Finger Style | Full |
For Use With (Application) | Critical Environments, Cleanrooms, and Electronics Assembly |
Glove Cuff Style | Knit |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction