Learn More
Wells Lamont™ Full-Finger Nylon Glove Liner Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Wells Lamont™ M555XL
 View more versions of this product
Catalog No. 19-048-646D
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Safety
No
Ambidextrous
Yes
X-Large
Gold
Machine wash in warm water with detergent or soap for 10-15 minutes. Water temperature should not exceed 60°C (140°F.)
Full
lining gloves
Nylon
White