Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Latex Palm

Fit and comfortable protection

Manufacturer: Wells Lamont Industry Group Y9282M

 View more versions of this product

Catalog No. 19148742

  Check Availability
Add to Cart

Alert

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


    Specifications

    Exterior Finish Seamless
    Glove Cuff Style Knit
    Glove Cuff Color Green
    For Use With (Application) ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
    Coated Area Palm, finger
    Color Gray
    Cut Resistance Rating Level 3
    Material Natural Foam Latex
    Size Medium

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit