Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Polyurethane Palm

Fit and comfortable protection

Manufacturer: Wells Lamont Industry Group Y9284XXL

 View more versions of this product

Catalog No. 19-148-832

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Seamless
    Glove Cuff Style Knit
    Glove Cuff Color Black
    For Use With (Application) protection from lacerations and superior resistance against cuts and for grip and abrasion resistance
    Material Polyurethane
    Size 2X-Large
    Coated Area Palm, finger
    Color Gray
    Cut Resistance Rating Level 3

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit