Wells Lamont™ GuardTec3™ Hi-Vis Foam Latex Palm Gloves

Manufacturer: Wells Lamont Industry Group Y9282HVL

 View more versions of this product

Catalog No. 17-200-308

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit wrist
    For Use With (Application) Cut protection, high visibility applications
    Cut Resistance Rating Level 3
    Exterior Finish Natural Foam Latex
    Glove Cuff Color Brown
    Material Nitrile Palm, FlexTech™ Back Material
    Size Large
    Thickness 13g
    Thumb Style Regular
    Coated Area Palm, finger
    Color Orange/Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit