Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ MechPro™ Synthetic Leather Palm Mechanics Gloves

Gloves are constructed of heavy duty, ribbed knit fabric for abrasion and incidental heat resistance. Specially designed and created for trade industries such as fleet mechanics, electricians, and millwrights.

$90.00

Specifications

Color Black
For Use With (Application) Fleet/Maintenance Mechanics, Pipe Fitters, Electricians, Millwrights, and Various Trades Industries
Glove Cuff Style Hook and Loop
Material Synthetic Leather
Product Type Specs Mechanics Gloves
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-130-5873
View Documents
Wells Lamont™
7701S
Small
Dozen of 12 for $90.00
Add to cart
 
19-130-5874
View Documents
Wells Lamont™
7701M
Medium
Dozen of 12 for $90.00
Add to cart
 
19-099-240C
View Documents
Wells Lamont™
7701L
Large
Dozen of 12 for $90.00
Add to cart
 
19-130-5876
View Documents
Wells Lamont™
7701XL
X-Large
Dozen of 12 for $90.00
Add to cart
 
Description

Description

  • Gloves provide grip and dexterity for handling tools or parts
  • Ribbed knit fabric for nick, abrasion, and incidental heat resistance
  • Synthetic leather palm and fingertips
  • Reinforced fingertips with pinched fingertip design
  • Gusseted finger crotch and perforated finger fourchettes for comfort
  • Double stitched in high wear areas: fingertips, index finger, and palm
  • Elastic cuff with Feeltite™ hook and loop adjustable wrist closure
Specifications

Specifications

Black
Hook and Loop
Mechanics Gloves
Ribbed Knit
Fleet/Maintenance Mechanics, Pipe Fitters, Electricians, Millwrights, and Various Trades Industries
Synthetic Leather
7701
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit