Wells Lamont™ Industrial Nylon Glove Liners

Ideal barrier between hand and latex glove

Manufacturer: Wells Lamont Industry Group M117M

 View more versions of this product

Catalog No. 19-044157

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Rinse all glove thoroughly in cold Water to ensure the release of particle in the product. If pre-soaking use luke warm Water to ensure good particle softening. Wash time should not be longer than 15 to 20 minute. Use a rinse cycle for the product in cold
    Glove Cuff Style Regular
    Hand Ambidextrous
    For Use With (Application) barrier between hand and latex gloves
    Laboratory/Cleanroom Use Yes
    Material Nylon
    Size Men's
    Color White
    Disposable No
    Finger Style Half
    Glove Cuff Color Black
    Sterile No

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit