Wells Lamont™ Industrial Scepter™ Polyester and Wire Cut-Resistant Glove Liners

Lightweight and comfortable

Manufacturer: Wells Lamont Industry Group M121XL

 View more versions of this product

Catalog No. 18-999-4858

  Check Availability
Add to Cart

    Specifications

    Hand Ambidextrous
    Hem Color Red
    For Use With (Application) cut and abrasion resistance
    Size X-Large
    Weight Light weight
    Coating Material Polyurethane
    Color White
    Cut Resistance Rating Level 4
    Material Stainless Steel, Polyester
    Sterile No

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit