Wells Lamont Kevlar w/Latex Coated Palm Gloves

Manufacturer: Wells Lamont Industry Group Y9245XL/DEL

 View more versions of this product

Catalog No. 19-148-703

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Cut Resistance Rating Level 2
    Glove Cuff Style Knit
    Glove Cuff Color Blue
    Hem Color Navy blue
    For Use With (Application) construction, recycling, lumber and mill works and landscaping
    Material Kevlar™
    Size X-Large
    Coating Material Seamless
    Color Yellow, blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit