Wells Lamont™ Safety King™ Hi-Vis Green Knit Palm-Coated Gloves

Machine knit gloves with brushed inside for comfort, dexterity and warmth

Manufacturer: Wells Lamont Industry Group SKG9295XL

 View more versions of this product

Catalog No. 19-148-628

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Glove Cuff Style Knit
    Glove Cuff Color Yellow
    For Use With (Application) road crews and general purposes
    Liner Material Unlined
    Size X-Large
    Coated Area Palm
    Color Hi-vis green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit