Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Scepter™ Sterile Antimicrobial M321 Cut-Resistant Gloves

Lightweight and sterile gloves protect hands from prolonged exposure to latex and have a high level of tactile sensation. Can be used as a liner or stand alone glove.

$775.50

Specifications

Certifications/Compliance ASTM F1790-97, ASTM F2992-15
Certifications/Compliance (Europe) Category III
Sterility Sterile
Color White
Cut Resistance Rating ANSI A4
View More Specs

Products 4
Catalog Number Mfr. No. Size Glove Cuff Color Price Quantity & Availability  
Catalog Number Mfr. No. Size Glove Cuff Color Price Quantity & Availability  
17-200-333
View Documents
Wells Lamont™
M321S
Small Green
Pack of 10 for $775.50
Case of 4 PK for $2,960.00
Add to cart
 
17-200-334
View Documents
Wells Lamont™
M321M
Medium Orange
Pack of 10 for $775.50
Case of 4 PK for $2,960.00
Add to cart
 
17-200-335
View Documents
Wells Lamont™
M321L
Large Blue
Pack of 10 for $775.50
Case of 4 PK for $2,960.00
Add to cart
 
17-200-336
View Documents
Wells Lamont™
M321XL
X-Large Red
Pack of 10 for $775.50
Case of 4 PK for $2,960.00
Add to cart
 
Description

Description

  • Stainless steel and polyester construction
  • Launderable 15 times without shrinking
  • Moderate tactile sensitivity
  • Antimicrobial
  • Sterile and ambidextrous

Applications: Medical, organ tissue processing and pharmaceutical

ANSI Cut Level 4

Specifications

Specifications

ASTM F1790-97, ASTM F2992-15
Sterile
ANSI A4
Category III
White
Stainless Steel, Polyester
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit