Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Scepter™ Sterile Antimicrobial M321 Cut-Resistant Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Stainless steel and polyester construction
- Launderable 15 times without shrinking
- Moderate tactile sensitivity
- Antimicrobial
- Sterile and ambidextrous
- Applications: Medical, organ tissue processing and pharmaceutical
- ANSI Cut Level 4
- Made in the USA
Specifications
Certifications/Compliance | ASTM F1790-97, ASTM F2992-15 |
Certifications/Compliance (Europe) | Category III |
Coating Material | Uncoated |
Color | White |
Cut Resistance Rating | ANSI A4 |
Hand | Ambidextrous |
Material | Stainless Steel Fiber, Polyester |
Product Type | Cut-resistant Gloves |
Size | Medium |
Sterility | Sterile |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction