Wells Lamont Sterile Spec-Tec Glove Liners

Cut resistant liners with tactile sensitivity

Manufacturer: Wells Lamont Industry Group M102L

 View more versions of this product

Catalog No. 19-048-682

  Check Availability
Add to Cart

    Specifications

    Certifications ASTM F 1790-97-CPPT
    Cleaning Procedure Laundered using a mild soap and Detergent, use a warm Water wash with a temperature not to exceed 160°F, wash time should not be longer than 15 to 20 min.
    Disposable No
    Finger Style Full
    Glove Cuff Style Knit
    Glove Cuff Color Blue
    Hand Ambidextrous
    Inspected for Medical Use Yes
    For Use With (Application) pathologists, coroners, hospital waste handlers and orthopedic, trauma and general surgeons and provides cut resistance
    Laboratory/Cleanroom Use Yes
    Material Polyethylene
    Size Large
    Sterile Yes
    Color White
    Cut Resistance Rating Level 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit