Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Cut Resistant Glove Liners

Catalog Number p-3203315
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight, thin glove liners are highly reusable and provide ANSI A1 cut resistance.

  • Lightweight, thin, and highly reusable white glove liner
  • Provides ANSI Cut Level A1 protection
  • Category II for particulates
  • Can be used alone or as a glove liner
  • Ambidextrous
  • Made in the USA
Color White
Cut Resistance Rating ANSI A1
Disposable Reusable
Hand Ambidextrous
Material HPPE
Product Type Cut-resistant Glove Liner
Size Small
For Use With (Application) Cleanroom, Medical, or Semi-Conductor Industries
Glove Cuff Style Knit
Hem Color White
Series M214
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit