Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Wells Lamont™ Premium Grain Cowhide Leather Palm Work Gloves

Gloves made of cow grain leather offer a full leather index finger, leather tips, and a leather knuckle strap. The lined palm provides additional comfort in the palm of the hand.

Specifications

Color Natural
Glove Cuff Style Gauntlet
Grade Premium
Material Premium Grain Cowhide Leather
Product Type Specs Work Gloves
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
11-462-32C
View Documents
Wells Lamont™
Y2009L
Large
Pack of 12 for $92.00
N/A
11-462-32D
View Documents
Wells Lamont™
Y2009XL
X-Large
Pack of 12 for $98.00
N/A
Description

Description

  • Premium quality grain cowhide
  • Grain knuckle strap
  • Heavy duty canvas back
  • Gauntlet cuff
  • Wing thumb
  • Gunn cut
Specifications

Specifications

Natural
Premium
Work Gloves
Gunn
Gauntlet
Premium Grain Cowhide Leather
Y2009
Wing
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit