Learn More
Wells Lamont™ Whizard™ Cut-Resistant Heavy-Duty Slipguard Gloves
Offer comfortable, heavy-duty cut resistance with slip protection
Manufacturer:  Wells Lamont™ 133801
 View more versions of this product
Catalog No. 11-400-29D
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Fabric consists of high-performance fibers and stainless-steel core to provide an excellent source of cut resistance
  • Polyurethane pattern covering palm, fingers and thumb crotch improves gripping
  • Ideal for the pulp and paper industry as well as for plastic trimming
  • Also features excellent thermal properties
  • 3 in. (7.6cm) ribbed cuffs
  • Overedge color indicates size: red (XXS), green (XS), yellow (S), blue (M), white (L), orange (XL)
Order Info
Hand-specific. Preshrunk and machine washable. Made in the USA.
Specifications
Specifications
Safety
Level 5
Hand-specific
the pulp and Paper industry, for Plastic trimming
Large
White
Polyurethane
Standard
White
Fiber, Stainless Steel
White