Wells Lamont™ Whizard™ Cut-Resistant Heavy-Duty Slipguard Gloves

Offer comfortable, heavy-duty cut resistance with slip protection

Manufacturer: Wells Lamont Industry Group 133556

 View more versions of this product

Catalog No. 11-400-27B

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) the pulp and Paper industry, for Plastic trimming
    Cut Resistance Rating Level 5
    Glove Cuff Style Continuous knit
    Glove Cuff Color Green
    Hand Hand-specific
    Hem Color Yellow
    Material Fiber, Stainless Steel
    Color Green
    Size Small
    Coating Material Polyurethane

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit