Wells Lamont™ Whizard™ Cut-Resistant Heavy-Duty Slipguard Gloves

Offer comfortable, heavy-duty cut resistance with slip protection

Manufacturer: Wells Lamont Industry Group 133800

 View more versions of this product

Catalog No. 11-400-28H

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Standard
    For Use With (Application) the pulp and Paper industry, for Plastic trimming
    Cut Resistance Rating Level 5
    Glove Cuff Color White
    Hand Right
    Hem Color White
    Material Fiber, Stainless Steel
    Size Large
    Coating Material Polyurethane
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit