Learn More
Wells Lamont MT130 Sterile/Non-Sterile Touchscreen Cut Resistant Gloves
Ultra-lightweight construction provides bare hand dexterity and superior tactile sensitivity
Manufacturer: Wells Lamont MT130L
Description
- Patent pending yarn technology delivers cut resistance and touchscreen capability
- Minimal particle shedding to meet cleanroom classifications and requirements
- Meets Category II using Helmke Drum Test referenced in IEST RP-CC003.3
- ANSI Level 4
- Ambidextrous gloves available in different sizes
Specifications
Large | |
Gray | |
Blue |