Learn More
Wells Lamont™ MT130 Sterile/Non-Sterile Touchscreen Cut Resistant Gloves
Ultra-lightweight construction provides bare hand dexterity and superior tactile sensitivity
Manufacturer:  Wells Lamont Industry Group MT130M
 View more versions of this product
Catalog No. 19-086-602
  Check Availability
Add to cart
Specifications
Description
  • Patent pending yarn technology delivers cut resistance and touchscreen capability
  • Minimal particle shedding to meet cleanroom classifications and requirements
  • Meets Category II using Helmke Drum Test referenced in IEST –RP-CC003.3
  • ANSI Level 4
  • Ambidextrous gloves available in different sizes
Description & Specifications
Specifications
Medium
Gray
Orange