Learn More
Wesco Industrial ProductsSupplier Diversity Partner Folding Handle Scissors Lift Table DFS Item
Manufacturer:  Wesco Industrial ProductsSupplier Diversity Partner 260201
Mobile Scissor Lift Table. Table top measures 17.75" Wide x 27.35" Long. Raised Heught 28.5". Lowered Height 8.5". Polyurethane Caster Size 4" x 1". Chrome plated handle and foot pedal. Lever release handle. Foot operated pump. Locking Swivel Casters. 98 lbs.
Catalog No. NC0953573