Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Polypropylene Disposable Beard Covers

Specifications

Color White
Material Polypropylene
Disposable Yes
For Use With (Application) Prevents beard and facial hair from contaminating food or getting caught in machinery
View More Specs

Products 2
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
19-068-442 View Documents West Chester
UBC1000
1000/Cs. Case of 1000 for $116.66
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-121-070 View Documents West Chester
UBC500
100/Cs. Pack of 100 for $35.21
Case of 5 PK for $174.91
N/A
Due to product restrictions, please to purchase or view availability for this product.
Specifications

Specifications

White
Polypropylene
Yes
Prevents beard and facial hair from contaminating food or getting caught in machinery
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit