West Chester PosiM3 Coveralls

Manufacturer: West Chester Marketing C3802/XXL

 View more versions of this product

Catalog No. 19-130-0531E

  Check Availability
Add to Cart

    Specifications

    Material Polypropylene
    Seam Type Serged
    Color White
    Size 2X-Large
    Size (English) Chest 29 in.
    Disposable Yes
    Size (Metric) Chest 73.9cm
    For Use With (Application) Industrial manufacturing, Paper industry, food processing or where heavy dust environments are present
    Size (English) Inseam 30.3 in.
    Size (Metric) Inseam 76.9cm
    Construction 5-ply SMMMS Technology
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit