Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester PosiM3 Coveralls

Catalog Number 191300531H
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Size (English) Chest:
24.4 in.
26 in.
27.6 in.
29 in.
31.5 in.
33.1 in.
34.6 in.
Size (English) Inseam:
28 in.
29 in.
29.5 in.
30.3 in.
31.5 in.
32.7 in.
33.5 in.
This item is not returnable. View return policy
This item is not returnable. View return policy
Size 5X-Large
Material Polypropylene
Color White
Disposable Single-use
Cuff Style Elastic
Seam Type Serged
Description 5-ply SMMMS Technology
Size (English) Chest 34.6 in.
Size (English) Inseam 33.5 in.
Size (Metric) Chest 87.8 cm
Size (Metric) Inseam 85 cm
For Use With (Application) Industrial manufacturing, Paper industry, food processing or where heavy dust environments are present
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit