Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester PosiUB Coveralls

Click to view available options
XX-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Size 2X-Large
Material Polyethylene, Polypropylene
Color White
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Seam Type Serged
Certifications/Compliance NFPA 99, U.S. ASTM 1670-1671
Description Attached hood, latex-free elastic wrist, ankle, waistband and face openings
For Use With (Application) Painting, asbestos abatement, remediation and Oil handling
Length (English) 41.3 in.
Length (English) Sleeve 35.8 in.
Length (Metric) 104.9 cm
Length (Metric) Sleeve 90.93 cm
Width (English) Chest 29 in.
Show More Show Less