West Chester Premium Blue Latex Gloves

Resist a wide range of chemicals and detergents

Manufacturer: West Chester Marketing 52L100/9

 View more versions of this product

Catalog No. 19-084836

  Check Availability
Add to Cart

Alert

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


    Specifications

    Certifications 21 CFR 170-199, USDA
    Exterior Finish Diamond
    Glove Cuff Style Pinked
    For Use With (Application) food service, janitorial, light assembly, agriculture, laboratories
    Size 9
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Natural Rubber
    Thickness 18mil
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit