Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Premium Top Grain Cowhide Leather Palm Gloves

Manufacturer:  West Chester Marketing 8000/S

 View more versions of this product

Catalog No. 19-821-562


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

11 x 4.25 in. (27.94 x 10.79cm)
Leather, cowhide
4.25 in.
White
27.94cm
Gauntlet
Small
10.79cm
11 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit