Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Reversible Jersey Gloves

$19.00 - $432.00

Specifications

For Use With (Application) manufacturing, general labor, material handling, shipping/receiving, agriculture
Glove Cuff Style Knit
Color Natural
Material Cotton
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-060004
View Documents
West Chester
KJ55I
Men's Dozen of 12 for $19.00
Add to cart
 
19-084375
View Documents
West Chester
KJ8I
Women's Case of 288 for $432.00
Add to cart
 
Specifications

Specifications

manufacturing, general labor, material handling, shipping/receiving, agriculture
Knit
Natural
Cotton
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit