Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Chester Safety Cuff Gloves
Click to view available options
Latex coated palm; Crinkle finish; Color: natural; Large
Description
- Protects against liquids
- Crinkle finish
- Safety Cuff added protection
- Coated palm and fingers
Specifications
Color | Natural |
Material | Latex |
Size | Large |
For Use With (Application) | Metal handling, general labor, maintenance |
Glove Cuff Style | Safety |
Liner Material | Jersey |
Lining | Lined |