Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Standard Split Cowhide Leather Palm Gloves with Rubberized Cuff

Gloves feature a rubberized safety cuff, cotton hem, and wing thumb

Specifications

Glove Cuff Style Safety
Color Gray
Grade Standard
Material Shoulder Split Cowhide Leather
Thumb Style Wing
View More Specs

Products 2
Catalog Number Mfr. No. Gender Price Quantity & Availability  
Catalog Number Mfr. No. Gender Price Quantity & Availability  
19-821-546
View Documents
West Chester™
500L
Men's This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-050-144
View Documents
West Chester™
500
Women's This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • 2.5 in. rubberized safety cuff
  • Standard split cowhide
  • Wing thumb
Specifications

Specifications

Safety
Standard
Wing
Gray
Shoulder Split Cowhide Leather
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit