West Chester Spunbond Polypropylene Coverall

Manufacturer: West Chester Marketing U1100/XL

 View more versions of this product

Catalog No. 19-821-592

  Check Availability
Add to Cart

    Specifications

    Material Spunbond Polypropylene
    Includes Open wrist, ankle
    Seam Type Serged
    Color White
    Size X-Large
    For Use With (Application) General labor, pharmaceuticals, cleaning, laboratory
    Closure Type Full Length Zipper Front
    Size (Metric) Inseam 73.66cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit