Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus Mouse anti-Virus, Unconjugated, Clone: E60, MilliporeSigma™
Mouse Monoclonal Antibody
$177.00 - $444.00
Specifications
Antigen | West Nile Virus |
---|---|
Clone | E60 |
Applications | Immunocytochemistry, Neutralization, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF22332MI
|
MilliporeSigma
MABF223325UG |
25 μg |
Each for $177.00
|
|
|||||
MABF22331MI
|
MilliporeSigma
MABF2233100UG |
100 μg |
Each for $444.00
|
|
|||||
Description
West Nile Virus Monoclonal antibody specifically detects West Nile Virus in Virus samples. It is validated for Immunocytochemistry, Neutralizing, Western Blotting.Specifications
West Nile Virus | |
Immunocytochemistry, Neutralization, Western Blot | |
Unconjugated | |
Mouse | |
Virus | |
E | |
IgG2a κ | |
Protein G Purified | |
Clone E60 is a mouse monoclonal antibody that specifically detects envelope protein of West Nile Virus. It targets an epitope within the DII fusion loop. |
E60 | |
Monoclonal | |
Purified | |
Epigenetics and Nuclear Function | |
E protein;Envelope protein;Env | |
West Nile Virus envelope protein. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. | |
∼47 kDa observed; 31.88 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title