Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
West Nile Virus Protein E Rabbit anti-Virus, DyLight 488, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305992G
Description
West Nile Virus Protein E Polyclonal antibody specifically detects West Nile Virus Protein E in Virus samples. It is validated for Western BlotSpecifications
West Nile Virus Protein E | |
Polyclonal | |
Western Blot | |
West Nile Virus envelope glycoprotein E | |
Produced in rabbits immunized with purified, recombinant West Nile Virus (WNV) (lineage 1, strain NY99) E/ Envelope (ABA62343.1; Gln586-Ala705). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Virus | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title