Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WFDC1, Goat anti-Human, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP280145

Catalog No. NB013296


Add to cart

Description

Description

WFDC1 Polyclonal antibody specifically detects WFDC1 in Human Samples. It is validated for Flow Cytometry, Peptide ELISA.
Specifications

Specifications

WFDC1
0.5 mg/mL
Flow Cytometry 10 ug/mL, Peptide ELISA Detection limit 1:32000
Affinity Purified
WFDC1
Peptide with sequence C-KNVAEPGRGQQKHFQ corresponding to C-Terminus according to NP_067020.2.
Immunogen affinity purified
RUO
Primary
58189
ELISA, Flow Cytometry
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
ps20 growth inhibitor, PS20Prostate stromal protein ps20, WAP four-disulfide core domain 1, WAP four-disulfide core domain 1 homolog, WAP four-disulfide core domain protein 1
Goat
IgG
0.1 mg
Store at -20°C. Avoid freeze/thaw cycles.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit