Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WFDC1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$482.50

Specifications

Antigen WFDC1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP156613
SDP
View Documents
Novus Biologicals
NBP156613
100 μL
Each of 1 for $482.50
Add to Cart
 
Description

Description

WFDC1 Polyclonal specifically detects WFDC1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

WFDC1
Polyclonal
Rabbit
Q9HC57
58189
Synthetic peptides corresponding to WFDC1(WAP four-disulfide core domain 1) Antibody(against the N terminal of WFDC1. Peptide sequence WKRALPARLAEKSRAEEAGAPGGPRQPRADRCPPPPRTLPPGACQAARCQ.
Primary
Western Blot
Unconjugated
RUO
ps20 growth inhibitor, PS20Prostate stromal protein ps20, WAP four-disulfide core domain 1, WAP four-disulfide core domain 1 homolog, WAP four-disulfide core domain protein 1
WFDC1
IgG
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit