Learn More
Aviva Systems Biology Corp WFDC5 Antibody - N-terminal region (ARP52854_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52854_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WFDC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WFDC5. Alternate names: PRG5, WAP1, dJ211D12.5ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Pig
Catalog No. 50-014-60852