Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WFDC5 Antibody (M47-P2D5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250363C
Description
WFDC5 Monoclonal specifically detects WFDC5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
WFDC5 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
WFDC5 | |
Ovalbumin-conjugated synthetic peptide TERVHDGRPG | |
0.1 mL | |
Cancer, Cellular Signaling | |
149708 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
M47-P2D5 | |
Western Blot, ELISA, Immunohistochemistry | |
dJ211D12.5, PRG5protease inhibitor WAP1, Putative protease inhibitor WAP1, WAP four-disulfide core domain 5, WAP four-disulfide core domain protein 5, WAP1p53-responsive gene 5 protein | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction