Learn More
Invitrogen™ WFDC5 Monoclonal Antibody (5G5)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA523092
Catalog No. PIMA523092
Only null left
Add to Cart
Description
Description
Peptide Sequence: RKKGEKSGGC PPDDGPCLLS VPDQCVEDSQ CPLTRKCCYR ACFRQCVPRV SVKLGSCPED QLRCLSPMNH LCHKDSDCSG KKRCCHSACG RDCRDPARG.
WFDC5 is a putative acid-stable proteinase inhibitor.This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. Most WFDC proteins contain only one WFDC domain, and this encoded protein contains two WFDC domains. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. Most WFDC gene members are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the centromeric cluster.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WFDC5
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8TCV5
WFDC5
WFDC5 (NP_663627, 25 a.a. ~ 123 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot
5G5
Unconjugated
WFDC5
BC019734; dJ211D12.5; p53-responsive gene 5 protein; Prg5; protease inhibitor WAP1; putative protease inhibitor WAP1; UNQ3036/PRO9842; WAP four-disulfide core domain 5; WAP four-disulfide core domain protein 5; WAP1; WFDC5
Mouse
Affinity chromatography
RUO
149708
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.