Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ 4 and 13mm Puradisc Polyethersulfone Syringe Filters
For minimal adsorptive protein loss
$582.00 - $1761.00
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Passes USP Class VI requirements for biosafety. |
Material | Polysulfone |
Sterility | Non-sterile |
For Use With (Application) | For filtering serum, plasma and tissue culture solutions – as well as other protein-containing solutions where minimal adsorptive protein loss is required. |
Catalog Number | Mfr. No. | Pore Size | Quantity | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Quantity | Sterility | Price | Quantity & Availability | |||
09-909-13
|
Cytiva
67821302 |
0.2 μm | 100/Pk. | Non-sterile |
Pack of 100 for $582.00
|
|
|||
09-909-10
|
Cytiva
67811304 |
0.45 μm | 500/Pk. | Non-sterile |
Pack of 500 for $1,761.00
|
|
|||
09-909-14
|
Cytiva
67821304 |
0.45 μm | 100/Pk. | Non-sterile |
Pack of 100 for $582.00
|
|
|||
Description
Suitable for filtering serum, plasma and tissue culture solutions—as well as other protein-containing solutions where minimal adsorptive protein loss is required.
- Polypropylene housing
- Autoclavable
- Female Luer lock on inlet side; male slip Luer on outlet side
4mm dia. Filters
- For samples up to 2mL
- Filter area: 0.2cm2 Hold-up volume (with air purge) <7.5μL
- For samples up to 10mL
- Filter area: 1.3cm2
- Hold-up volume (after air purge) <25μL
Specifications
Autoclavable | |
Polysulfone | |
For filtering serum, plasma and tissue culture solutions – as well as other protein-containing solutions where minimal adsorptive protein loss is required. | |
Female Luer Lok | |
Polypropylene | |
≤10 mL |
Passes USP Class VI requirements for biosafety. | |
Non-sterile | |
≤25 μL | |
Male Luer Slip | |
Syringe Filter | |
13 mm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title