Learn More
Cytiva Whatman™ 8-Channel Filter Plates
For use with Agilent 850-DS dissolution sampling station
Supplier: Cytiva 77073200
Description
- 0.45μm membrane pore size
- Available in PTFE, Nylon, PES, and GMF filter media
- Disposable
Automated dissolution testing
Specifications
Agilent 850-DS Dissolution Sampling Station | |
PES | |
0.45 μm | |
Automated dissolution testing |