Learn More
Cytiva (Formerly GE Healthcare Life Sciences) 8-Channel Filter Plates
For use with Agilent 850-DS dissolution sampling station
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 77073400
 View more versions of this product
Catalog No. 09-924-000
Add to cart
Description
Description
  • 0.2µm membrane pore size
  • Available in PTFE, Nylon, PES, and GMF filter media
  • Disposable
Automated dissolution testing
Specifications
Specifications
0.2 μm
Automated dissolution testing
Agilent 850-DS sampling station
PTFE