Learn More
Cytiva (Formerly GE Healthcare Life Sciences) 8-Channel Filter Plates Available on GSA/VA Contract for Federal Government customers only.
For use with Agilent 850-DS dissolution sampling station
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 77073600
 View more versions of this product
Catalog No. 09-924-002
Add to cart
Description
Description
  • 0.45µm membrane pore size
  • Available in PTFE, Nylon, PES, and GMF filter media
  • Disposable
Automated dissolution testing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Automated dissolution testing
Agilent 850-DS sampling station
PES
0.2 μm