Learn More
Cytiva (Formerly GE Healthcare Life Sciences) 8-Channel Filter Plates Available on GSA/VA Contract for Federal Government customers only.
For use with Agilent 850-DS dissolution sampling station
$2070.00
Specifications
For Use With (Application) Automated dissolution testing
For Use With (Equipment) Agilent 850-DS sampling station
View More Specs
Products 7
Catalog Number Mfr. No. Material Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Material Pore Size Price Quantity & Availability  
09-929-000
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
77073000
PTFE 0.45 μm Pack of 50 for $2,070.00
Add to cart
09-929-001
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
77073100
Nylon 0.45 μm Pack of 50 for $2,070.00
Add to cart
09-929-003
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
77073300
GMF 0.45 μm Pack of 50 for $2,070.00
Add to cart
09-929-002
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
77073200
PES 0.45 μm Pack of 50 for $2,070.00
Add to cart
09-924-000
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
77073400
PTFE 0.2 μm Pack of 50 for $2,070.00
Add to cart
09-924-001 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
77073500
Nylon 0.2 μm Pack of 50 for $2,070.00
Add to cart
09-924-002 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
77073600
PES 0.2 μm Pack of 50 for $2,070.00
Add to cart
Description
Description
  • 0.45µm membrane pore size
  • Available in PTFE, Nylon, PES, and GMF filter media
  • Disposable
Automated dissolution testing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Automated dissolution testing
Agilent 850-DS sampling station