Learn More
Cytiva Whatman™ 90 mm Nuclepore™ Polycarbonate Track-Etched Membranes PROMO
Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.
Specifications
Diameter (Metric) 90 mm
Flow Rate High
For Use With (Application) Epifluorescence microscopy, environmental analysis, cell biology
Material Nucleopore Polycarbonate
View More Specs
Products 2
Catalog Number Mfr. No. Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Price Quantity & Availability  
09-920-022LR0
View Documents
Cytiva
29489294
0.2 μm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
09-920-023LR0
View Documents
Cytiva
29489295
0.4 μm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Effective chemotaxis requires a delicate balance between cell adhesion and migration. Nucleopore Membranes combine PVP-free polypropylene with sharp, uniform cylindrical pores to achieve both.
  • Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
Specifications
Specifications
90 mm
High
Epifluorescence microscopy, environmental analysis, cell biology
Nucleopore Polycarbonate