Whatman™ 903 Neonatal Screening Cards
Manufacturer: Whatman™ 10537279
 View more versions of this product
Catalog No. 09-800-964
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Quantity Each circle holds 75 to 80uL of blood
    For Use With (Application) Specimen collection
    Material Cellulose
    Product Type 903 Multiple-part neonatal card
  • Description & Specifications
Related Products
Product Content Feedback