Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman 96-Well Lysate Clarification Plate
Ideal for the removal of cellular debris from lysates during high-throughput plasmid DNA purification
$999.00
Catalog Number | Mfr. No. | Volume (Metric) Well | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) Well | Includes | Price | Quantity & Availability | ||||
09-003-8
|
Cytiva (Formerly GE Healthcare Life Sciences)
77700062 |
Long drip director |
Pack of 25 for $999.00
|
|
Description
- Polystyrene plate with a proprietary filter membrane
- Processes 96 lysates in <10 min.
- Compatible with vacuum filtration or centrifugation
- Produces recovery rates 10 to 30% higher than manual centrifuge methods
- Recovers an average of 6µg DNA/well when used with the Whatman DNA Binding Plate (Cat. No. PF77002810)
- Adaptable to automated platforms