Learn More
GE Healthcare Whatman™ Accessories for Mini-UniPrep™ Syringeless Filters
Used with Mini-UniPrep syringeless plunger filter units for fast and easy HPLC/UHPLC sample preparation. Whatman™ Compressors for Mini-UniPrep™ Syringeless Filters process one or multiple filter vials simultaneously for
Manufacturer:  GE Healthcare CR0000001
 View more versions of this product
Catalog No. 09-923-15
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Whatman Mini-UniPrep Syringeless Filters integrate an autosampler vial, filtration membrane, plunger, and cap/septa into one consumable product. Compressors activate the plunger to filter the sample through the membrane without hand stress.
  • Compatible with Mini-UniPrep and/or Mini-UniPrep G2 syringeless filter vials
  • Multi-chamber compressors process multiple vials simultaneously for stream-lined filtration
  • Mini-UniPrep filter vials prepare samples 3x faster than standard syringe filters, and can be loaded directly into an autosampler
  • Options include: 1-, 6- or 8-position hand or handle compressors (additional 8-position trays also available)
Specifications
Specifications
Mini-UniPrep syringeless filters
Sample filtration/preparation for HPLC/UHPLC
Single-sample Hand Compressor; One position
Compressor
Single-Sample Handle Compressor
Polypropylene
One sample vial
1Each