Learn More
GE Healthcare Whatman™ Accessories for Mini-UniPrep™ Syringeless Filters
Used with Mini-UniPrep syringeless plunger filter units for fast and easy HPLC/UHPLC sample preparation. Whatman™ Compressors for Mini-UniPrep™ Syringeless Filters process one or multiple filter vials simultaneously for
Manufacturer:  GE Healthcare MUPG2HCPWC1
 View more versions of this product
Catalog No. 09-924-490
Add to cart
Description
Description
Whatman Mini-UniPrep Syringeless Filters integrate an autosampler vial, filtration membrane, plunger, and cap/septa into one consumable product. Compressors activate the plunger to filter the sample through the membrane without hand stress.
  • Compatible with Mini-UniPrep and/or Mini-UniPrep G2 syringeless filter vials
  • Multi-chamber compressors process multiple vials simultaneously for stream-lined filtration
  • Mini-UniPrep filter vials prepare samples 3x faster than standard syringe filters, and can be loaded directly into an autosampler
  • Options include: 1-, 6- or 8-position hand or handle compressors (additional 8-position trays also available)
Specifications
Specifications
Mini-UniPrep G2 syringeless filters
Sample filtration/preparation for HPLC/UHPLC
Purple/White
One sample vial
1Each
Single-sample Compressor
Polypropylene
Hand compressor, Single position (Promo)
Compressor