Whatman™ Acid/Alkali Test Papers

Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.

Manufacturer: Whatman™ 2638500

 View more versions of this product

Catalog No. 09-800-978

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Format Strips
    Quantity 10 books of 20 strips
    Product Type pH Test Paper
    Color Indicator Congo Red
    pH Range 6.8 to 8.3

    • Litmus Blue turns red in acid
    • Litmus Red turns blue in base, at pH 5 to 8
    • Congo Red ph range: 6.8 to 8.3

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit