Learn More
GE Healthcare Whatman™ Acid-Treated Low-Metal TCLP Filters Available on GSA/VA Contract for Federal Government customers only.
Binder free filters
Manufacturer:  GE Healthcare 1810150
 View more versions of this product
Catalog No. 09-922-2
Add to cart
Description
Description
The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6µm to 0.8µm
  • Resistant to acids and bases
  • Available in six diameters, including sizes to fit Büchner funnels and zero-headspace extraction units
  • Pretreatment with acid eliminates need to acid-wash filters in your lab, for reduced analysis time and reagent costs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
Conform to requirements of U.S. EPA Method 1311 Toxicity Characteristic Leaching Procedure (TCLP), including 0.6 to 0.8µm particle retention requirement.
Specifications
Specifications
GE’s Whatman TCLP test filters are acid-treated with low metal content. They support TCLP testing by providing accurate information about TCLP limits to ensure the best possible disposal plan can be put into action.
150 mm
0.7 μm
60 mL/min
Determine the leaching potential in a landfill for hazardous organic and inorganic contaminants that could potentially migrate into groundwater, threatening drinking water sources
TCLP Limits Test Filter, 150 mm circle (50 pcs)
5.9 in.
Circles
420 μm
15cm
Borosilicate Glass